missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement (100X)

Catalog No. a4000046402
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000046402 100 mL
A4000046401 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4000046402 Supplier Gibco™ Supplier No. A4000046402
Only null left

Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement is an animal-origin-free (AOF) basal medium supplement that is used to reduce the amount of FBS needed to culture cells.

Gibco Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement is an animal-origin-free (AOF) basal medium supplement. The insulin and transferrin in the ITS-X Select formulation are produced in non-mammalian recombinant expression systems.

ITS-X is used as a basal medium supplement to reduce the amount of fetal bovine serum (FBS) used to culture cells. It is routinely used, in combination with 2–4% FBS, to support cell types that require ethanolamine, such as bronchial epithelial cells.

Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids. Transferrin is an iron carrier and may also help reduce toxic levels of oxygen radicals and peroxide. Selenium, as sodium selenite, is a co-factor for glutathione peroxidase and other proteins that is used as an antioxidant in media. Ethanolamine is a precursor of phospho-glycerides, which are essential to the structure of the plasma membrane and cellular organelles.

The product formulation contains the following components:

  • Insulin (1000.0 mg/L)
  • Transferrin (550.0 mg/L)
  • Sodium selenite (0.67 mg/L)
  • Ethanolamine (200.0 mg/L)

cGMP manufacturing and quality system

ITS-X Select Supplement is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Endotoxin Level ≤ 1.00 EU/mL
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 275 to 310 mOsm/kg
pH 7.8 to 9.5
Quantity 100 mL
Shipping Condition Room Temperature
Source Animal Origin-free
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.