missing translation for 'onlineSavingsMsg'
Learn More

Instrument Check Standards for ICP/MS, Set of 5, SPEX CertiPrep™

Catalog No. CLICSSET
Change view
Click to view available options
Quantity:
5 x 125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLICSSET 5 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLICSSET Supplier SPEX CertiPrep Group LLC Supplier No. CLICSSET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Instrument Check Standards for ICP-MS, 125 mL, Set of 5, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Instrument Check Standards for ICP-MS
  • Set of 5
  • 125 mL Volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Instrument Check Standards, Set of 5
Packaging Triple-leached LDPE bottle
Name Note Set of 5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Quantity 5 x 125 mL
Grade ICP/ICP-MS
Concentration or Composition (by Analyte or Components) Set contains one 100mL bottle each of:, CL-ICS-1, CLHG2-1AY, CL-ICS-3, CL-ICS-4, CL-ICS-5
Show More Show Less

Product intended for laboratory use only