missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Instrument Check Standard 8 For Environmental Analysis, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Al | 50μg/mL |
| As | 50μg/mL |
| Co | 50μg/mL |
| Cr | 50μg/mL |
| Cu | 50μg/mL |
| K | 50μg/mL |
| Na | 50μg/mL |
| P | 50μg/mL |
| Pb | 50μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | ICP Instrument Check Standard 8 |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | 2% HNO3 |
| Quantity | 125 mL |
| Grade | Environmental |
| Concentration or Composition (by Analyte or Components) | Each component at 50μg/mL in 2% HNO3:, Aluminum, Arsenic, Chromium, Cobalt, Copper, Lead, Phosphorus, Potassium, Sodium |
Product intended for laboratory use only