missing translation for 'onlineSavingsMsg'
Learn More

Instrument Calibration Standard 2 For CLP, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ICAL2

Catalog No. ICAL-2


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ni400μg/mL
Zn200μg/mL
Mn150μg/mL
Ag100μg/mL
Cr100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Instrument Calibration Standard 2
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
5% HNO3
Components in 5% HNO3:
Cr    •100μg/mL
Mn    •150μg/mL
Ni    •400μg/mL
Ag    •100μg/mL
Zn    •200μg/mL
Triple-leached LDPE bottle
GC-MS
Certified Reference Material
125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only