missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INSR beta Monoclonal Antibody (CT-1)
GREENER_CHOICE

Catalog No. PIMA513778
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513778 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513778 Supplier Invitrogen™ Supplier No. MA513778
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA5-13778 targets Insulin Receptor beta in ELISA, AP, FACS, ICC/IF and IP applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13778 immunogen is a 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor beta-subunit.

Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.

Specifications

Antigen INSR beta
Applications Affinity Purification, ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CT-1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene INSR
Gene Accession No. P06213, P15127, P15208
Gene Alias 4932439J01Rik; CD220; CD220 beta; CD221; D630014A15Rik; HHF5; IGF-I receptor; Insr; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR beta; IR-A; IR-B
Gene Symbols INSR
Host Species Mouse
Immunogen A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor beta-subunit (aa 1368-1382).
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16337, 24954, 3643
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less