missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ InSolution™ Sinefungin, Calbiochem™,

Catalog No. 5670512MG
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5670512MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5670512MG Supplier MilliporeSigma™ Supplier No. 5670512MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended Storage

−20°C

Chemical Identifiers

CAS 58944-73-3
Molecular Formula C15H23N7O5
Molecular Weight (g/mol) 381.393
InChI Key LMXOHSDXUQEUSF-LWCRYBRRSA-N
Synonym insolution sinefungin, 2,5-diamino-6-2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl hexanoic acid, antibiotic 32232rp, compound-57926
PubChem CID 6917665
IUPAC Name 2,5-diamino-6-[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]hexanoic acid
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)CC(CCC(C(=O)O)N)N)O)O

Specifications

Packaging Glass Bottle
Quantity 2 mg
Physical Form Liquid
Recommended Storage −20°C