missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Inserts for 2 mL Large Opening Vials

Insert fits 12 x 32 mm vials with large opening (6 mm)

Supplier:  MilliporeSigma™ Supelco™ 27407

 View more versions of this product

Catalog No. 111010523


Only null left

Description

Description

  • Glass conical inserts with polymer feet are used to contain standard mixtures for various experimental purposes.
Specifications

Specifications

Glass
Step Vials
100
0.25 mL
Flange
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions