missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Flakes, Technical, Spectrum™ Chemical

Catalog No. 18606563
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606563 50 kg Fiber Drum
18606560 500 g Poly Bottle
18606561 2.5 kg Poly Bottle
18606562 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606563 Supplier Spectrum Chemical Mfg Cor Supplier No. S130850KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1308, 1310-73-2, NaOH

Spectrum™ Chemical Sodium Hydroxide, Flakes, Technical is a highly caustic metallic base combined with alkali salt. Commonly known as Caustic soda, this inorganic compound is water soluble, as well as in ethanol and methanol. The dissolution of solid Sodium Hydroxide in water creates a highly exothermic reaction (creates a lot of heat). One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Used in many industries, most notably in the manufacture of pulp, paper, textiles, drinking water, detergents and as a pharmaceutical excipient. Technical grade Sodum Hydroxide products are suitable for general laboratory use.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 1
UN Number UN1823
Quantity 50 kg
Grade Technical
Packaging Fiber Drum
Chemical Name or Material Sodium Hydroxide