missing translation for 'onlineSavingsMsg'
Learn More

Inositol, NF, 97-102%, Spectrum™ Chemical

Catalog No. p-7157608
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603452 100 g Poly Bottle
18603453 500 g Poly Bottle
18603454 2.5 kg Poly Bottle
18603455 12 kg Poly Pail
18603456 25 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603452 Supplier Spectrum Chemical Mfg Cor Supplier No. I1012100GM
Only null left

I1012, 87-89-8, C6H12O6

Spectrum™ Chemical Inositol, NF

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES OC1C(O)C(O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 100 g
Percent Purity 97 to 102%
Grade NF