missing translation for 'onlineSavingsMsg'
Learn More

Inositol, FCC, 97%, Spectrum™ Chemical

Catalog No. p-7157605
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603447 100 g Poly Bottle
18603448 500 g Poly Bottle
18603449 2.5 kg Poly Pail
18603450 12 kg Poly Pail
18603451 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603447 Supplier Spectrum Chemical Mfg Cor Supplier No. I1004100GM
Only null left

I1004, 87-89-8, C6H12O6

Spectrum™ Chemical Inositol, FCC is a vitamin-like substance, sometimes known colloquially as ′vitamin B-8′, is a beneficial nutrient that has been implicated in the treatment of some behavioral or emotional disorders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES OC1C(O)C(O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range 224-227 C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 97%
Grade FCC