missing translation for 'onlineSavingsMsg'
Learn More

Inositol, 97%, Spectrum™ Chemical

Catalog No. 18611042
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611042 50 kg Poly Bottle
18611039 100 g Poly Bottle
18611043 500 g Poly Bottle
18611041 2.5 kg Poly Bottle
18611040 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611042 Supplier Spectrum Chemical Mfg Cor Supplier No. IN14050KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES OC1C(O)C(O)C(O)C(O)C1O

Specifications

CAS Max % 1
Melting Point 224° to 227°C
Packaging Poly Bottle
Quantity 50 kg
Percent Purity 0.97
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Inositol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.