missing translation for 'onlineSavingsMsg'
Learn More

Inosine Pranobex 98.0+%, TCI America™

Catalog No. I10371G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10371G 1 g
I1037100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10371G Supplier TCI America Supplier No. I10371G
Only null left

Chemical Identifiers

CAS 36703-88-5
Molecular Formula C52H78N10O17
Molecular Weight (g/mol) 1115.249
MDL Number MFCD05662374
InChI Key YLDCUKJMEKGGFI-QCSRICIXSA-N
PubChem CID 37510
IUPAC Name 4-acetamidobenzoic acid;9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;1-(dimethylamino)propan-2-ol
SMILES CC(CN(C)C)O.CC(CN(C)C)O.CC(CN(C)C)O.CC(=O)NC1=CC=C(C=C1)C(=O)O.CC(=O)NC1=CC=C(C=C1)C(=O)O.CC(=O)NC1=CC=C(C=C1)C(=O)O.C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)CO)O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 1115.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Inosine Pranobex