Learn More
Thermo Scientific Chemicals Inosine-5'-monophosphate disodium salt hydrate
CAS: 352195-40-5 | C10H11N4Na2O8P | 392.17 g/mol
Supplier: Thermo Scientific Chemicals J6195922
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Inosine-5'-monophosphate disodium salt hydrate | |
| White | |
| MFCD00036201,MFCD00036201 | |
| disodium 5'-inosinate | |
| [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O | |
| 392.17 | |
| 392.17 (Anhydrous) |
| 352195-40-5 | |
| C10H11N4Na2O8P | |
| 100 g | |
| AANLCWYVVNBGEE-IDIVVRGQSA-L | |
| [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate;sodium | |
| 123134061 | |
| Crystalline Powder |
Chemical Identifiers
| 352195-40-5 | |
| 392.17 | |
| AANLCWYVVNBGEE-IDIVVRGQSA-L | |
| 123134061 | |
| [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O |
| C10H11N4Na2O8P | |
| MFCD00036201,MFCD00036201 | |
| disodium 5'-inosinate | |
| [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate;sodium |
Safety and Handling
EINECSNumber : 225-146-4
RTECSNumber : NM7519500
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only