Learn More
MilliporeSigma™ InnoZyme™ TACE Activity Kit
Catalog No. CBA0421KIT
Change view
Click to view available options
Quantity:
1kit
Catalog No. Quantity
CBA0421KIT 1kit
1 options
Catalog No. CBA0421KIT Supplier MilliporeSigma™ Supplier No. CBA0421KIT
Only null left
Designed to measure human TACE activity in cell lysates and biological samples and for screening enzyme inhibitors
  • Highly specific: based on immunocapture of human TACE
  • Highly sensitive and quantitative: assay range 5 to 100ng/mL
  • Convenient: 96-well format and non-radioactive detection
  • Versatile: appropriate for measuring active TACE and high-throughput screening of TACE inhibitors
Specifications
Product Type TACE Activity Kit
Kit Contents Anti-Human TACE-Coated 96-Well Plate, Control, Substrate, Sample Buffer, Assay Buffer, Wash Buffer, Plate Sealer, and a user protocol.
Quantity 1kit
Storage Requirements -20°C
For Research Use Only