missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ InnoZyme™ Myeloperoxidase Activity Kit
Description
- Uses polyclonal antibody specific for human MPO immobilized on 96-well plate to specifically capture enzyme
- Activity of captured myeloperoxidase is measured using detection reagent that includes TMB and hydrogen peroxide
- Following color development, reaction is stopped with sulfuric acid and absorbance of oxidized TMB is detected at 450nm
Specifications
Specifications
| Product Type | Myeloperoxidase Activity Kit |
| Kit Contents | Anti-MPO Coated 96-Well Plate, MPO Standard, TMB, Hydrogen Peroxide, Assay Buffer, Sample Buffer, Stop Solution, Plate Sealers, and a user protocol. |
| Quantity | 1kit |
| Storage Requirements | -20°C |
For Research Use Only