missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 20 OD
High efficiency antibody binding for maximum sensitivity
$3014.53 - $3014.53
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
|---|---|
| Target | 40nm Colloidal Gold Nanoparticles 20 OD |
| Product Type | Antibody Labeling Kit |
Alert:
For Research Use Only
Description
- Uniform Shape
- Low co-efficient of variation in terms of size
- High efficiency antibody binding for maximum sensitivity
- Available at high concentrations to simplify assay development
Specifications
| Store at 4°C. Do not freeze. | |
| 40nm Colloidal Gold Nanoparticles 20 OD | |
| Antibody Labeling Kit |