missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Inkjet Vinyl Label
Make an impact with this inkjet-printable vinyl label outfitted with a specially formulated topcoat that invigorates colors and label appearance.
Supplier: Brady™ J203892595
Description
- Flexible material withstands harsh elements, including liquids, alcohol, chemicals, and indoor fluorescent and UV lighting
- Topcoat also protects against smudges, smears and splotches
- An ink cartridge is required to print this material
Chemical Identifiers
| Not Autoclavable | |
| None | |
| Matte | |
| Single-side Printable | |
| Label | |
| Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking | |
| -40°C to +60°C | |
| 0.137 mm |
| Permanent Acrylic | |
| White | |
| Vinyl | |
| Ink Jet | |
| Non-sterile | |
| -40°C | |
| 0.0054 in. |
Specifications
| Not Autoclavable | |
| None | |
| Matte | |
| Vinyl | |
| Ink Jet | |
| Non-sterile | |
| J2000 Printer | |
| 500 Labels | |
| -40°F to +140°F | |
| 0.0054 in. | |
| 1.5 x 1.5 in. | |
| 1.5 in. | |
| 38.1 mm |
| Permanent Acrylic | |
| White | |
| Die-cut | |
| Single-side Printable | |
| Label | |
| Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking | |
| -40°C | |
| Round | |
| -40°C to +60°C | |
| 0.137 mm | |
| 1.5 in.. | |
| 1.5 in. | |
| 38.1 mm |