Learn More
Brady™ Inkjet Vinyl Label
Make an impact with this inkjet-printable vinyl label outfitted with a specially formulated topcoat that invigorates colors and label appearance.
Supplier:  Brady™ J403882595
 View more versions of this product
Catalog No. 17377040
Only null left
Description
Description
  • Flexible material withstands harsh elements, including liquids, alcohol, chemicals, and indoor fluorescent and UV lighting
  • Topcoat also protects against smudges, smears and splotches
  • An ink cartridge is required to print this material
Specifications
Chemical Identifiers
Not Autoclavable
None
Matte
Single-side Printable
Label
Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking
-40°C to +60°C
0.137 mm
Permanent Acrylic
White
Vinyl
Ink Jet
Non-sterile
-40°C
0.0054 in.
Specifications
Not Autoclavable
None
Matte
Vinyl
Ink Jet
Non-sterile
J4000 Printer
500 Labels
-40°F to +140°F
0.0054 in.
1 x 1 in.
1 in.
25.4 mm
Permanent Acrylic
White
Die-cut
Single-side Printable
Label
Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking
-40°C
Round
-40°C to +60°C
0.137 mm
1 in..
1 in.
25.4 mm
Product Suggestions
Product Suggestions