missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Inkjet Polyester Label and Overlaminate Kit
Take your inkjet printing further and label faster with these ideally-sized label and overlaminate kits.
Supplier: Brady™ J202612569KT
Description
- Just peel, adhere and apply with these pre-sized labels and overlaminate
- Inkjet-printable label is outfitted with a specially formulated topcoat that invigorates colors and label appearance
- Label topcoat also protects against smudges, smears and splotches
- Acrylic overlaminate provides up to 10 years of outdoor durability through improved UV and abrasion resistance
- Overlaminate is oversized by a quarter-inch for easier application and complete coverage
- An ink cartridge is required to print label material
Chemical Identifiers
| Not Autoclavable | |
| None | |
| Matte | |
| Polyester | |
| Non-sterile | |
| 2-roll Kit | |
| Rectangle | |
| -40°C to +60°C | |
| 0.1143 mm |
| Permanent Acrylic | |
| White | |
| Die-cut | |
| Ink Jet | |
| Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking | |
| -40°C | |
| -40°F to +140°F | |
| 0.0045 in. |
Specifications
| Not Autoclavable | |
| None | |
| Matte | |
| Polyester | |
| Ink Jet | |
| Non-sterile | |
| J2000 Printer | |
| -40°C | |
| Rectangle | |
| -40°F to +140°F | |
| 0.0045 in. | |
| 4.33 x 2.95 in. | |
| 109.98 mm | |
| 74.93 mm |
| Permanent Acrylic | |
| White | |
| Die-cut | |
| Single-side Printable | |
| Label | |
| Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking | |
| 2-roll Kit | |
| 270 Labels | |
| 109.98 x 74.93 mm | |
| -40°C to +60°C | |
| 0.1143 mm | |
| 4.33 in. | |
| 2.95 in. |