missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ InkJet Microscope Slides

Catalog No. 22042921
Change view
Click to view available options
Color:
Aqua
Blue
Burnt Orange
Green
Lilac
Orange
Pink
Tan
White
Yellow
Corner Style:
45°
90°
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Corner Style
22042921 Orange 45°
12550102 Green 90°
12550106 Burnt Orange 90°
12550103 Pink 90°
12550100 White 90°
22042922 Lilac 45°
22042915 White 45°
22042916 Blue 45°
22042923 Tan 45°
12550107 Lilac 90°
22042918 Pink 45°
22042917 Green 45°
22042920 Aqua 45°
12550105 Aqua 90°
22042919 Yellow 45°
12550101 Blue 90°
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22042921 Supplier Fisherbrand™ Supplier No. IJL61064600621
Only null left

Superfrost™/ColorFrost™ microscope slides designed for use with Leica™ Microsystems IP S inkjet printer

  • Printing instruments use chemical-resistant ink and print directly on Superfrost/ColorFrost tab of the slide
  • For histology, cytology, hematology, and microbiology applications
  • Clipped 45° or straight 90° corners
  • Frosted tabs in nine different colors

Specifications

Frosted Tabs
Color Orange
Corner Style 45°