Learn More
Thermo Scientific™ Injection Port Liners with CarboFrit for Thermo Scientific™ Trace and Focus SSL Instruments
Catalog No. 03151604
Change view
Click to view available options
For Use With (Equipment):
Split Straight Liner
Catalog No. For Use With (Equipment)
03151604 Split Straight Liner
1 options
Catalog No. 03151604 Supplier Thermo Scientific™ Supplier No. 453T2131
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments.
Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments. Highly deactivated and produced to exacting tolerances.
Specifications
Product Type Split Straight Liner
Quantity 5
Length (Metric) 105 mm
Packing Material CarboFrit
Description 453T2131
Diameter (Metric) Inner 5 mm
Diameter (Metric) Outer 8 mm
For Use With (Equipment) Split Straight Liner