missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Initial Calibration Verification Standard IA For CLP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ICV1A
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ca | 5000μg/mL |
| K | 5000μg/mL |
| Mg | 5000μg/mL |
| Na | 5000μg/mL |
| Al | 200μg/mL |
| Ba | 200μg/mL |
| Fe | 100μg/mL |
| Sb | 60μg/mL |
| Co | 50μg/mL |
| V | 50μg/mL |
| Ni | 40μg/mL |
| Cu | 25μg/mL |
| Zn | 20μg/mL |
| Mn | 15μg/mL |
| Ag | 10μg/mL |
| As | 10μg/mL |
| Cr | 10μg/mL |
| Tl | 10μg/mL |
| Be | 5μg/mL |
| Cd | 5μg/mL |
| Se | 5μg/mL |
| Pb | 3μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Initial Calibration Verification Standard 1A | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride | |
| Components in 5% HNO3/tr. Tartaric Acid/tr. HF: Al •200μg/mL Sb •60μg/mL As •10μg/mL Ba •200μg/mL Be •5μg/mL Cd •5μg/mL Ca •5000μg/mL Cr •10μg/mL Co •50μg/mL Cu •25μg/mL Fe •100μg/mL Pb •3μg/mL Mg •5000μg/mL Mn •15μg/mL Ni •40μg/mL K •5000μg/mL Se •5μg/mL Ag •10μg/mL Na •5000μg/mL Tl •10μg/mL V •50μg/mL Zn •20μg/mL |
| Triple-leached LDPE bottle | |
| GC-MS | |
| Certified Reference Material | |
| 500 mL |
Product intended for laboratory use only