missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Infusorial Earth (Kieselguhr) (Diatomaceous Silica), Reagents
Infusorial Earth (Kieselguhr) (Diatomaceous Silica), Technical, for general laboratory use. Manufactured in ISO 9001 facility.
Chemical Identifiers
| CAS | 61790-53-2 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| Synonym | Kieselguhr, Amorphous silicon dioxide |
| IUPAC Name | silanedione |
| SMILES | O=[Si]=O |
Chemical Identifiers
| 61790-53-2 | |
| 60.08 | |
| Kieselguhr, Amorphous silicon dioxide | |
| O=[Si]=O |
| O2Si | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione |
Specifications
| Solid | |
| 2230°C | |
| O2Si | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| Infusorial Earth (Kieselguhr) (Diatomaceous Silica) |
| 61790-53-2 | |
| Poly Pail | |
| Kieselguhr, Amorphous silicon dioxide | |
| O=[Si]=O | |
| 60.08 |