missing translation for 'onlineSavingsMsg'
Learn More

Infusorial Earth (Kieselguhr) (Diatomaceous Silica), Reagents

Catalog No. 01337213
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337213 2.5 kg
01337212 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337213 Supplier Labchem Inc Supplier No. C22170002.5KG
Only null left

Infusorial Earth (Kieselguhr) (Diatomaceous Silica), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 61790-53-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Kieselguhr, Amorphous silicon dioxide
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Solid
Quantity 2.5 kg
Boiling Point 2230°C
Packaging Poly Pail
Chemical Name or Material Infusorial Earth (Kieselguhr) (Diatomaceous Silica)