missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza B NP Monoclonal Antibody (H89B), FITC
GREENER_CHOICE

Catalog No. MA17306
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17306 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17306 Supplier Invitrogen™ Supplier No. MA17306
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

The MA1-7306 antibody reacts with Influenza B virus nuclear protein. MA1-7306 has been successfully used in immunofluorescence and immunocytochemistry applications with acetone fixation. This antibody does not work well in Western blotting applications. The MA1-7306 antibody was raised against the Influenza B Nucleoprotein.

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.

Specifications

Antigen Influenza B NP
Applications Immunocytochemistry
Classification Monoclonal
Clone H89B
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias Influenza B; Influenza NP
Host Species Mouse
Immunogen Influenza B Nucleoprotein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less