missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza B NP Monoclonal Antibody (H89B)

Description
MA1-7058 detects the Influenza B virus. This antibody is specific for a nuclear protein. MA1-7058 has been successfully used in immunofluorescence procedures. The MA1-7058 immunogen is Influenza B virus lysate.
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
Specifications
Specifications
| Antigen | Influenza B NP |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | H89B |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Influenza B; Influenza NP |
| Host Species | Mouse |
| Immunogen | Influenza B virus lysate. |
| Show More |