missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza B NA Monoclonal Antibody (B3)
GREENER_CHOICE

Catalog No. 01671851
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01671851 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01671851 Supplier Invitrogen™ Supplier No. MA110823
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA1-10823 detects Influenza B Neuraminidase. MA1-10823 has been successfully used in ELISA and inhibition applications. The MA1-10823 immunogen is Influenza B Lee/40.

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.

Specifications

Antigen Influenza B NA
Applications ELISA, Inhibition Assays
Classification Monoclonal
Clone B3
Concentration 0.80 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Influenza NA
Host Species Mouse
Immunogen Influenza B Lee/40.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less