missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza A H5N1 HA Chimeric Recombinant Rabbit Monoclonal Antibody (310-18A3)
GREENER_CHOICE

Catalog No. MA551687
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551687 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551687 Supplier Invitrogen™ Supplier No. MA551687
Only null left

Rabbit Recombinant Monoclonal Antibody

This antibody is specific for the H5N1 and H1N1 subtypes of influenza A virus. Influenza viruses are negative-sense, single-stranded, segmented RNA viruses.

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.

Specifications

Antigen Influenza A H5N1 HA Chimeric
Applications ELISA, Functional Assay
Classification Recombinant Monoclonal
Clone 310-18A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Rabbit
Immunogen This antibody was raised by cloning antibody sequences form B cells isolated from people immunized with the H5 DNA vaccine.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less