missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec Inflammatory Bowel Disease Control Slides
Normal Human Tissue Inflammatory Bowel Disease Control Slides, with Ileum Or General Intestine.
$2,216.21 - $4,466.32
Specifications
| Type | Control Slide |
|---|---|
| For Use With (Application) | Use as a Control |
| Content And Storage | Room-Temperature |
Description
- High-quality control slides designed to assist in accurate histopathology and laboratory testing
- Includes well-preserved normal human tissue to support reliable diagnostic comparisons
- Easy-to-use slides suitable for laboratory and educational purposes
- Prepared to maintain tissue integrity for consistent experimental results
Specifications
| Control Slide | |
| Use as a Control | |
| Room-Temperature |