missing translation for 'onlineSavingsMsg'
Learn More

Mopec Inflammatory Bowel Disease Control Slides

Catalog No. 21010274
Change view
Click to view available options
Quantity:
100 Control Slides
25 Control Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21010274 25 Control Slides
21010273 100 Control Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21010274 Supplier Mopec Supplier No. SC95725
Only null left

Normal Human Tissue Inflammatory Bowel Disease Control Slides, with Ileum Or General Intestine.

  • High-quality control slides designed to assist in accurate histopathology and laboratory testing
  • Includes well-preserved normal human tissue to support reliable diagnostic comparisons
  • Easy-to-use slides suitable for laboratory and educational purposes
  • Prepared to maintain tissue integrity for consistent experimental results

Specifications

Content And Storage Room-Temperature
Includes 24 Unstained 1 Stained Control Slide
Quantity 25 Control Slides
Type Control Slide
For Use With (Application) Use as a Control