Learn More
Synthware™ Inert Gas/Vacuum Double Bank Manifold
GREENER_CHOICE
Catalog No. 31501742
Change view
Click to view available options
Port Size (Metric):
5 ports
Catalog No. Port Size (Metric)
31501742 5 ports
1 options
Catalog No. 31501742 Supplier Synthware™ Supplier No. M471005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0 to 4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Front Left, Rear Left and Right Hose Connection
Specifications
Type Vacuum/Inert Gas Double Manifold
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Port Size (Metric) 5 ports
Width (Metric) 550 mm