Learn More
3M™ Maintenance Sorbent Pads M-A2002
Description
- Sorbent pads designed for use in and around the work area to catch leaks and drips
- For use on most non-aggressive liquids
- Very easy to use and to retrieve
- Promotes a cleaner, safer working environment
Order Info
dimProductType
Specifications
Specifications
| Color | Grey |
| Type | Sorbent Pads |
| Length (Metric) | 520 mm |
| Width (Metric) | 400 mm |