missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl Acetate 98.0+%, TCI America™

Catalog No. p-7116695
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A00681G 1 g
A00685G 5 g
2 options

Catalog No. A00681G

Supplier: TCI America A00681G

Only null left

Chemical Identifiers

CAS 608-08-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00014561
InChI Key JBOPQACSHPPKEP-UHFFFAOYSA-N
Synonym 3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
PubChem CID 11841
IUPAC Name 1H-indol-3-yl acetate
SMILES CC(=O)OC1=CNC2=CC=CC=C21

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Indoxyl Acetate
RTECSNumber AI3325000
TSCA Yes