missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl acetate, 97%

Catalog No. AC156660050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156660050 5 g Glass bottle
AC156660010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156660050 Supplier Thermo Scientific Chemicals Supplier No. 156660050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 608-08-2 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-08-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00014561
InChI Key JBOPQACSHPPKEP-UHFFFAOYSA-N
Synonym 3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
PubChem CID 11841
IUPAC Name 1H-indol-3-yl acetate
SMILES CC(=O)OC1=CNC2=CC=CC=C21

Specifications

Melting Point 128.0°C to 131.0°C
Color Colorless to Lilac
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 71
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,chloroform
Formula Weight 175.19
Percent Purity 97%
Physical Form Glistening Crystals
Chemical Name or Material Indoxyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 210-154-2

RUO – Research Use Only