missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Indoline-2-carboxylic Acid 97.0+%, TCI America™
Supplier: TCI America I039425G
Specifications
| (+/-)-Indoline-2-carboxylic Acid | |
| C9H8NO2 | |
| 25 g | |
| QNRXNRGSOJZINA-QMMMGPOBSA-M | |
| (2S)-2,3-dihydro-1H-indole-2-carboxylate | |
| 86074 | |
| ≥97.0% (HPLC,T) |
| 78348-24-0 | |
| MFCD00010635 | |
| indoline-2-carboxylic acid, +/--indoline-2-carboxylic acid, dl-indoline-2-carboxylic acid, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 2-indolinecarboxylic acid, indoline-2-carboxylicacid, 1-indoline-2-carboxylic acid, pubchem3035 | |
| [O-]C(=O)[C@@H]1CC2=CC=CC=C2N1 | |
| 162.17 | |
| 163.18 | |
| Crystalline Powder |
Chemical Identifiers
| 78348-24-0 | |
| 162.17 | |
| QNRXNRGSOJZINA-QMMMGPOBSA-M | |
| 86074 | |
| [O-]C(=O)[C@@H]1CC2=CC=CC=C2N1 |
| C9H8NO2 | |
| MFCD00010635 | |
| indoline-2-carboxylic acid, +/--indoline-2-carboxylic acid, dl-indoline-2-carboxylic acid, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 2-indolinecarboxylic acid, indoline-2-carboxylicacid, 1-indoline-2-carboxylic acid, pubchem3035 | |
| (2S)-2,3-dihydro-1H-indole-2-carboxylate |
Safety and Handling
TSCA : No