missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Indoline-2-carboxylic Acid 97.0+%, TCI America™

Catalog No. p-7131382
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03945G 5 g
I039425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03945G Supplier TCI America Supplier No. I03945G
Only null left

Chemical Identifiers

CAS 78348-24-0
Molecular Formula C9H8NO2
Molecular Weight (g/mol) 162.17
MDL Number MFCD00010635
InChI Key QNRXNRGSOJZINA-QMMMGPOBSA-M
Synonym indoline-2-carboxylic acid, +/--indoline-2-carboxylic acid, dl-indoline-2-carboxylic acid, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 2-indolinecarboxylic acid, indoline-2-carboxylicacid, 1-indoline-2-carboxylic acid, pubchem3035
PubChem CID 86074
IUPAC Name (2S)-2,3-dihydro-1H-indole-2-carboxylate
SMILES [O-]C(=O)[C@@H]1CC2=CC=CC=C2N1

Specifications

Quantity 5 g
Formula Weight 163.18
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Indoline-2-carboxylic Acid