missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole 99.0+%, TCI America™
$46.20 - $460.11
Chemical Identifiers
| CAS | 120-72-9 |
|---|---|
| Molecular Formula | C8H7N |
| Molecular Weight (g/mol) | 117.151 |
| MDL Number | MFCD00005607 |
| InChI Key | SIKJAQJRHWYJAI-UHFFFAOYSA-N |
| Synonym | indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german |
| PubChem CID | 798 |
| ChEBI | CHEBI:16881 |
| IUPAC Name | 1H-indole |
| SMILES | C1=CC=C2C(=C1)C=CN2 |
Chemical Identifiers
| 120-72-9 | |
| 117.151 | |
| SIKJAQJRHWYJAI-UHFFFAOYSA-N | |
| 798 | |
| 1H-indole |
| C8H7N | |
| MFCD00005607 | |
| indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german | |
| CHEBI:16881 | |
| C1=CC=C2C(=C1)C=CN2 |
Specifications
| 120-72-9 | |
| White | |
| C8H7N | |
| 25 g | |
| indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german | |
| C1=CC=C2C(=C1)C=CN2 | |
| 117.151 | |
| CHEBI:16881 | |
| ≥99.0% (GC) | |
| Indole |
| 53°C | |
| 253°C | |
| MFCD00005607 | |
| 2811 | |
| SIKJAQJRHWYJAI-UHFFFAOYSA-N | |
| 1H-indole | |
| 798 | |
| 117.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0130
RTECSNumber : NL2450000
TSCA : Yes