missing translation for 'onlineSavingsMsg'
Learn More

Indole 99.0+%, TCI America™

Catalog No. I0021500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0021500G 500 g
I002125G 25 g
I0021100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0021500G Supplier TCI America Supplier No. I0021500G

Chemical Identifiers

CAS 120-72-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00005607
InChI Key SIKJAQJRHWYJAI-UHFFFAOYSA-N
Synonym indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
PubChem CID 798
ChEBI CHEBI:16881
IUPAC Name 1H-indole
SMILES C1=CC=C2C(=C1)C=CN2

Specifications

Melting Point 53°C
Color White
Boiling Point 253°C
Quantity 500 g
UN Number 2811
Formula Weight 117.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole
EINECSNumber (5)-0130
RTECSNumber NL2450000
TSCA Yes