missing translation for 'onlineSavingsMsg'
Learn More

Indole, 99%

CAS: 120-72-9 | C8H7N | 117.151 g/mol

Supplier:  Thermo Scientific Chemicals A144270B

 View more versions of this product

Catalog No. AAA144270B


Only null left
Explore available promotions

Description

Description

Indole is used in the preparation of gramine, which is obtained by Mannich reaction dimethylamine and formaldehyde. It is used as a starting material for the preparation of oxindole and indole-3-carboxaldehyde. In Diels-Alder reaction, it acts as dienophile and reacts with 2-aminofuran to get strychnine. It is the active component of jasmine oil, neroli oil and orange-blossom oil used in the perfume industry. Further, it is involved in the preparation of synthetic jasmine oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole is used in the preparation of gramine, which is obtained by Mannich reaction dimethylamine and formaldehyde. It is used as a starting material for the preparation of oxindole and indole-3-carboxaldehyde. In Diels-Alder reaction, it acts as dienophile and reacts with 2-aminofuran to get strychnine. It is the active component of jasmine oil, neroli oil and orange-blossom oil used in the perfume industry. Further, it is involved in the preparation of synthetic jasmine oil.

Solubility
Soluble in methanol and water.

Notes
Light and air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, iron and iron salts. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Indole
50°C to 54°C
6
121°C (249°F)
C8H7N
1000 g
107693
14,4963
Soluble in methanol and water.
C1=CC=C2C(=C1)C=CN2
117.151
CHEBI:16881
99%
120-72-9
1.22
253°C to 254°C
Unpleasant
MFCD00005607
UN2811
Light sensitive
indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
SIKJAQJRHWYJAI-UHFFFAOYSA-N
1H-indole
798
117.15

Chemical Identifiers

120-72-9
117.151
SIKJAQJRHWYJAI-UHFFFAOYSA-N
798
1H-indole
C8H7N
MFCD00005607
indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
CHEBI:16881
C1=CC=C2C(=C1)C=CN2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H318-H302
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P333+P313-P361-P363-P501c

H302-H311-H317-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 204-420-7

RTECSNumber : NL2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only