missing translation for 'onlineSavingsMsg'
Learn More

Indole-7-carboxylic Acid 97.0+%, TCI America™

Catalog No. I06061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06061G Supplier TCI America Supplier No. I06061G

Chemical Identifiers

CAS 1670-83-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210442
InChI Key IPDOBVFESNNYEE-UHFFFAOYSA-N
Synonym indole-7-carboxylic acid, 7-indolecarboxylic acid, indole-7-carboxylicacid, zlchem 444, pubchem2475, 7-carboxyindole, acmc-209dvl, indole-7-carboxlic acid, indole 7-carboxylic acid, 1h-indole-7-carboxylicacid
PubChem CID 74281
IUPAC Name 1H-indole-7-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)NC=C2

Specifications

Melting Point 205°C
Quantity 1 g
Formula Weight 161.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Indole-7-carboxylic Acid