missing translation for 'onlineSavingsMsg'
Learn More

Indole-7-carboxylic acid, 97%

Catalog No. p-7036611
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5493103 1 g
AAH5493106 5 g
AAH5493114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5493103 Supplier Thermo Scientific Chemicals Supplier No. H5493103

CAS: 1670-83-3 | C9H7NO2 | 161.16 g/mol

Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1670-83-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210442
InChI Key IPDOBVFESNNYEE-UHFFFAOYSA-N
Synonym indole-7-carboxylic acid, 7-indolecarboxylic acid, indole-7-carboxylicacid, zlchem 444, pubchem2475, 7-carboxyindole, acmc-209dvl, indole-7-carboxlic acid, indole 7-carboxylic acid, 1h-indole-7-carboxylicacid
PubChem CID 74281
IUPAC Name 1H-indole-7-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)NC=C2

Specifications

Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 161.05
Percent Purity 97%
Chemical Name or Material Indole-7-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-801-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only