missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 455540010

 View more versions of this product

Catalog No. AC455540010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Indole-6-methanol
Authentic
Glass Bottle
MFCD02179595
6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol
OCC1=CC=C2C=CNC2=C1
147.18
147.17
1075-26-9
96% min. (GC)
C9H9NO
1g
WRMZOPANDOHWJU-UHFFFAOYSA-N
(1H-indol-6-yl)methanol
2773459
97%

Chemical Identifiers

1075-26-9
147.18
WRMZOPANDOHWJU-UHFFFAOYSA-N
2773459
OCC1=CC=C2C=CNC2=C1
C9H9NO
MFCD02179595
6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol
(1H-indol-6-yl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only