missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-methanol, 97%, Thermo Scientific™

Catalog No. p-7077561
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455542500 250mg
AC455540010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455542500 Supplier Thermo Scientific Chemicals Supplier No. 455542500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1075-26-9
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD02179595
InChI Key WRMZOPANDOHWJU-UHFFFAOYSA-N
Synonym 6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol
PubChem CID 2773459
IUPAC Name 1H-indol-6-ylmethanol
SMILES OCC1=CC=C2C=CNC2=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 250mg
Formula Weight 147.17
Percent Purity 97%
Chemical Name or Material Indole-6-methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only