missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7131574
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
I07431G 1 g
I07435G 5 g
2 options

Catalog No. I07431G

Supplier: TCI America I07431G

Only null left

Chemical Identifiers

CAS 1196-70-9
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD02179596
InChI Key VSPBWOAEHQDXRD-UHFFFAOYSA-N
Synonym indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole
PubChem CID 2773435
IUPAC Name 1H-indole-6-carbaldehyde
SMILES C1=CC(=CC2=C1C=CN2)C=O

Specifications

Melting Point 129°C
Quantity 1 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole-6-carboxaldehyde