accessibility menu, dialog, popup

UserName

CAS RN 1196-70-9

IUPAC Name: 1H-indole-6-carbaldehyde
Synonyms: 1H-indole-6-carbaldehyde
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: VSPBWOAEHQDXRD-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2C=CNC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)