missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-4-methanol, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Formula Weight | 147.17 |
| Percent Purity | 97% |
| Chemical Name or Material | Indole-4-methanol |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present a |
RUO – Research Use Only