accessibility menu, dialog, popup

UserName

HOHN

CAS RN 1074-85-7

IUPAC Name: (1H-indol-4-yl)methanol
Synonyms: 1H-indol-4-ylmethanol
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: BVSGXWCTWBZFEV-UHFFFAOYSA-N
SMILES: OCC1=C2C=CNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)