missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-propionic acid, 98%

Catalog No. p-7044149
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0487706 5 g
AAL0487714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0487706 Supplier Thermo Scientific Chemicals Supplier No. L0487706
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 830-96-6 | C11H11NO2 | 189.21 g/mol

Indole-3-propionic acid is a reactant for preparation of fluorescent analogues of strigolactones, melanocortin receptors ligands, histamine H4 receptor agonists, and NR2B/NMDA receptor antagonists. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used to Study as an adjunct to improve perfusion after liver transplant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-propionic acid is a reactant for preparation of fluorescent analogues of strigolactones, melanocortin receptors ligands, histamine H4 receptor agonists, and NR2B/NMDA receptor antagonists. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used to Study as an adjunct to improve perfusion after liver transplant.

Solubility
It is slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 830-96-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005660
InChI Key GOLXRNDWAUTYKT-UHFFFAOYSA-N
Synonym 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
PubChem CID 3744
ChEBI CHEBI:43580
IUPAC Name 3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)CCC1=CNC2=CC=CC=C12

Specifications

Melting Point 132°C to 135°C
Quantity 5 g
Beilstein 147733
Solubility Information It is slightly soluble in water.
Formula Weight 189.21
Percent Purity 98%
Chemical Name or Material Indole-3-propionic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-600-1
RTECSNumber NM1329000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only