missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7131166
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00285G 5 g
I002825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00285G Supplier TCI America Supplier No. I00285G
Only null left

Chemical Identifiers

CAS 771-50-6
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005624
InChI Key KMAKOBLIOCQGJP-UHFFFAOYSA-N
Synonym indole-3-carboxylic acid, 3-indoleformic acid, 3-carboxyindole, indole-3-carboxylicacid, 3-indolecarboxylic acid, indole-3-carboxylate, 3-indolylcarboxylic acid, beta-indolylcarboxylic acid, indole-3 carboxylic acid, ico
PubChem CID 69867
ChEBI CHEBI:24809
IUPAC Name 1H-indole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CN2)C(=O)O

Specifications

Melting Point 234°C
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Indole-3-carboxylic Acid
EINECSNumber (5)-6656
RTECSNumber NK7882892
TSCA No