missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carbonitrile, 98%

Catalog No. AAA1604103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1604103 1 g
AAA1604106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1604103 Supplier Thermo Scientific Chemicals Supplier No. A1604103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5457-28-3 | C9H6N2 | 142.16 g/mol

Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 5457-28-3
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00022717
InChI Key CHIFTAQVXHNVRW-UHFFFAOYSA-N
Synonym 3-cyanoindole, indole-3-carbonitrile, 3-indolecarbonitrile, chembl46724, zlchem 360, 3-cyano-1h-indole, pubchem7325, acmc-209lhi, 1h-indol-3-yl cyanide
PubChem CID 230282
IUPAC Name 1H-indole-3-carbonitrile
SMILES N#CC1=CNC2=CC=CC=C12

Specifications

Melting Point 177°C to 182°C
Odor Odorless
Quantity 1 g
Beilstein 120888
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 142.16
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material Indole-3-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement

H302-H315-H319-H335

Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only